siRNA Details
Gene Symbol: Tmem104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033393.2 | NP_001028565.1 | 8 | 799 | ||
GenBank mRNA | AK040894.1 | - | 8 | 757 | ||
AK049868.1 | - | 8 | 736 | |||
AK131112.1 | - | 8 | 776 | |||
AK155041.1 | - | 8 | 776 | |||
AK167098.1 | - | 8 | 755 | |||
AK171314.1 | - | 8 | 776 | |||
AK171457.1 | - | 8 | 761 | |||
BC140976.1 | - | 8 | 719 | 9055588 |
