siRNA Details
Gene Symbol: Ubn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177185.4 | NP_796159.3 | 12 | 2205 | ||
GenBank mRNA | AK051609.1 | - | 12 | 1578 | ||
BC043095.1 | - | 1450 | ||||
BC044744.1 | - | 7 | 907 | |||
BC051458.1 | - | 12 | 2180 | 6468017 |
