siRNA Details
Gene Symbol: D5Ertd577e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177187.4 | NP_796161.3 | 3 | 878 | ||
GenBank mRNA | AK054417.1 | - | 3 | 878 | ||
BC053383.1 | - | 2 | 787 | |||
BC094905.1 | - | 2 | 775 | 30047197 | ||
BC099941.1 | - | 3 | 862 | 30031929 | ||
BC145804.1 | - | 3 | 824 | 40131107 |
Gene Symbol: E330014E10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122668.1 | NP_001116140.1 | 3 | 998 | ||
GenBank mRNA | AK087741.1 | - | 3 | 1000 |
Gene Symbol: Gm3286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122678.2 | NP_001116150.1 | 3 | 876 | ||
NM_001167793.1 | NP_001161265.1 | 3 | 916 | |||
GenBank mRNA | AK145737.1 | - | 5 | 1172 | ||
DV649213.1 | - | 1 | 267 | |||
DV653222.1 | - | 584 | ||||
DV655246.1 | - | 66 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
