siRNA Details
Gene Symbol: Tmem88b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033394.3 | NP_001028566.2 | 2 | 1642 | ||
GenBank mRNA | AK031860.1 | - | 1 | 61 | ||
AK035998.1 | - | 2 | 1642 | |||
AK038855.1 | - | 547 | ||||
AK158459.1 | - | 2 | 1643 |
