siRNA Details
Gene Symbol: Zfp629 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177226.5 | NP_796200.1 | 3 | 2878 | ||
GenBank mRNA | AK031161.1 | - | 3 | 2587 | ||
AK034484.1 | - | 3 | 2550 | |||
AK172933.1 | - | 4 | 2608 | |||
BC035547.1 | - | 3 | 2520 | 4481313 | ||
BC067053.1 | - | 3 | 2878 | 30543652 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
