siRNA Details
Gene Symbol: Fam19a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177233.5 | NP_796207.2 | 3 | 486 | ||
GenBank mRNA | AK081531.1 | - | 3 | 469 | ||
AK149355.1 | - | 3 | 489 | |||
AY325123.1 | - | 3 | 269 | |||
BB642822.1 | - | 3 | 486 | |||
BC089490.1 | - | 3 | 446 | 30504398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
