siRNA Details
Gene Symbol: Vstm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178791.4 | NP_848906.1 | 8 | 1003 | ||
GenBank mRNA | AK033244.1 | - | 8 | 959 | ||
AK040699.1 | - | 8 | 851 | |||
AK053685.1 | - | 8 | 1004 | |||
BC107254.2 | - | 8 | 550 | |||
BC125012.1 | - | 8 | 549 | |||
BC153828.1 | - | 8 | 550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
