siRNA Details
Gene Symbol: Pkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199593.1 | NP_001186522.1 | 4 | 610 | ||
NM_177262.4 | NP_796236.2 | 4 | 721 | |||
GenBank mRNA | AK089431.1 | - | 4 | 585 | ||
AK134792.1 | - | 5 | 705 | |||
AK149649.1 | - | 4 | 584 | |||
AK150062.1 | - | 4 | 547 | |||
AK170556.1 | - | 4 | 678 | |||
AK171297.1 | - | 4 | 593 | |||
BC052923.1 | - | 4 | 668 | 6312892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
