siRNA Details
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146043.2 | NP_001139515.1 | 6 | 519 | ||
NM_001271463.1 | NP_001258392.1 | 6 | 519 | |||
NM_001271464.1 | NP_001258393.1 | 6 | 519 | |||
NM_001271465.1 | NP_001258394.1 | 6 | 546 | |||
NM_177266.5 | NP_796240.3 | 6 | 552 | |||
GenBank mRNA | AK028450.1 | - | 6 | 505 | ||
AK035819.1 | - | 6 | 458 | |||
AK044371.1 | - | 6 | 520 | |||
AK083441.1 | - | 6 | 484 | |||
AK161797.1 | - | 6 | 455 | |||
AK165871.1 | - | 6 | 513 | |||
AK219489.1 | - | 5 | 365 | |||
BC027341.1 | - | 6 | 503 | 3984129 | ||
BC138233.1 | - | 6 | 450 | 8861252 | ||
BC145208.1 | - | 6 | 450 | 9053759 | ||
BY742160.1 | - | 6 | 520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
