siRNA Details
Gene Symbol: C530008M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163793.1 | NP_001157265.1 | 6 | 2639 | ||
GenBank mRNA | AK030106.1 | - | 6 | 2641 | ||
AK122469.1 | - | 1 | 812 | |||
BC086807.1 | - | 1 | 1258 |
