siRNA Details
Gene Symbol: C530008M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163793.1 | NP_001157265.1 | 6 | 2319 | ||
GenBank mRNA | AK030106.1 | - | 6 | 2321 | ||
AK122469.1 | - | 1 | 492 | |||
BC086807.1 | - | 1 | 938 |

More Information
Other Species Targeted

Gene Ontology Categories:
