siRNA Details
Gene Symbol: C530008M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163793.1 | NP_001157265.1 | 6 | 3428 | ||
GenBank mRNA | AK029850.1 | - | 1 | 502 | ||
AK030106.1 | - | 6 | 3430 | |||
AK042228.1 | - | 1 | 400 | |||
AK049629.1 | - | 1 | 638 | |||
AK050000.1 | - | 1 | 203 | |||
AK122469.1 | - | 1 | 1601 | |||
BC086807.1 | - | 1 | 2047 |
