siRNA Details
Gene Symbol: Sirpb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002898.1 | NP_001002898.1 | 2 | 132 | ||
GenBank mRNA | AB109404.1 | - | 209 | |||
AB112022.1 | - | 2 | 132 | |||
AB112023.1 | - | 135 | ||||
AB112024.1 | - | 169 | ||||
AK036935.1 | - | 127 | ||||
AK040982.1 | - | 185 | ||||
AK041165.1 | - | 189 | ||||
AK054545.1 | - | 145 | ||||
AK170671.1 | - | 135 | ||||
BC145417.1 | - | 144 | 9053975 | |||
BC145418.1 | - | 145 | 9053976 |
Gene Symbol: LOC100038947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173459.2 | NP_001166930.1 | 2 | 184 | ||
GenBank mRNA | AK155731.1 | - | 185 | |||
BC039636.1 | - | 128 | ||||
DQ055453.1 | - | 2 | 133 |
Gene Symbol: Sirpb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173460.1 | NP_001166931.1 | 2 | 225 | ||
GenBank mRNA | AK154879.1 | - | 2 | 226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
