siRNA Details
Gene Symbol: Cdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009865.2 | NP_033995.1 | 12 | 2456 | ||
GenBank mRNA | AF183946.1 | - | 12 | 2225 | ||
AK138606.1 | - | 12 | 2458 | |||
AK161927.1 | - | 1 | 1497 | |||
BC052056.1 | - | 12 | 2164 | 5696194 | ||
BC055808.1 | - | 3 | 1027 | |||
BC062962.1 | - | 12 | 2164 | 5696194 |
