siRNA Details
Gene Symbol: Zfp605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163996.1 | NP_001157468.1 | 4 | 545 | ||
GenBank mRNA | AK043719.1 | - | 4 | 546 | ||
AK051473.1 | - | 5 | 660 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163996.1 | NP_001157468.1 | 4 | 545 | ||
GenBank mRNA | AK043719.1 | - | 4 | 546 | ||
AK051473.1 | - | 5 | 660 |