siRNA Details
Gene Symbol: Mical2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193305.1 | NP_001180234.1 | 23 | 3295 | ||
NM_177282.5 | NP_796256.1 | 18 | 2614 | |||
GenBank mRNA | AK030608.1 | - | 18 | 2595 | ||
AK053370.1 | - | 11 | 3796 | |||
AK138935.1 | - | 18 | 2588 | |||
AK143191.1 | - | 18 | 2561 | |||
AK161075.1 | - | 18 | 2561 | |||
AK163078.1 | - | 18 | 2581 | |||
AK220353.1 | - | 21 | 2972 | |||
BC111895.1 | - | 16 | 2338 | 40061129 | ||
BC112415.1 | - | 16 | 2338 | 40061128 |
