siRNA Details
Gene Symbol: Ccbe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178793.4 | NP_848908.1 | 4, 5 | 480 | ||
GenBank mRNA | AK028377.1 | - | 4, 5 | 478 | ||
AK035153.1 | - | 4, 5 | 475 | |||
AK039742.1 | - | 4, 5 | 410 | |||
AK087673.1 | - | 498 | ||||
AK220435.1 | - | 4, 5 | 450 | |||
BC103803.1 | - | 4, 5 | 430 | 30088062 | ||
BC152322.1 | - | 4, 5 | 449 | 40140631 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
