siRNA Details
Gene Symbol: Cyp4f39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177307.3 | NP_796281.1 | 7, 8 | 857 | ||
GenBank mRNA | AK028950.1 | - | 7, 8 | 858 | ||
AK037192.1 | - | 7, 8 | 858 | |||
BC145755.1 | - | 7, 8 | 854 | 40131058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
