siRNA Details
Gene Symbol: Xpnpep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177310.2 | NP_796284.1 | 3 | 336 | ||
GenBank mRNA | AK042110.1 | - | 4 | 523 | ||
AK088324.1 | - | 3 | 336 | |||
BC137569.1 | - | 3 | 320 | 8860588 | ||
BC144717.1 | - | 3 | 321 | 9053265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
