siRNA Details
Gene Symbol: 6330408A02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177312.4 | NP_796286.2 | 2 | 154 | ||
GenBank mRNA | AK018141.1 | - | 2 | 117 | ||
AK028865.1 | - | 2 | 103 | |||
AK044247.1 | - | 1 | 1086 | |||
AK132923.1 | - | 2 | 156 | |||
AK160064.1 | - | 1 | 793 | |||
AK162673.1 | - | 2 | 125 | |||
BC075681.1 | - | 1 | 128 | 30608389 | ||
CV783961.1 | - | 2 | 155 |
