siRNA Details
Gene Symbol: Upf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081132.1 | NP_001074601.1 | 3 | 1185 | ||
GenBank mRNA | BC138358.1 | - | 3 | 1148 | 8861378 | |
BC138359.1 | - | 3 | 1148 | 8861379 |

More Information
Other Species Targeted

Gene Ontology Categories:
