siRNA Details
Gene Symbol: Tnfsf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177371.3 | NP_796345.3 | 3, 4 | 389 | ||
GenBank mRNA | AF520786.1 | - | 3, 4 | 306 | ||
AY764130.1 | - | 3, 4 | 389 | |||
BC096494.1 | - | 3, 4 | 375 | 30740802 | ||
BC138207.1 | - | 3, 4 | 354 | 8861226 | ||
BC138208.1 | - | 3, 4 | 354 | 8861227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
