siRNA Details
Gene Symbol: PWP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005049.2 | NP_005040.2 | 2, 3 | 213 | ||
XM_011529667.2 | XP_011527969.1 | 2, 3 | 278 | |||
GenBank mRNA | AK314184.1 | - | 2, 3 | 217 | ||
BC013309.2 | - | 2, 3 | 123 | 3528790 | ||
BC014988.1 | - | 2, 3 | 123 | 4906611 | ||
BM835826.1 | - | 2, 3 | 217 | |||
U53346.1 | - | 2, 3 | 147 | |||
U56085.1 | - | 2, 3 | 158 | |||
X95263.1 | - | 2, 3 | 154 |
Gene Symbol: LOC102724159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006723910.1 | XP_006723973.1 | 2, 3 | 250 |
