siRNA Details
Gene Symbol: Xaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037713.4 | NP_001032802.2 | 3 | 351 | ||
GenBank mRNA | AK140315.1 | - | 3 | 316 | ||
CK130151.1 | - | 2 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
