siRNA Details
Gene Symbol: Slc39a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026244.2 | NP_080520.2 | 6 | 1278 | ||
GenBank mRNA | AK014732.1 | - | 6 | 984 | ||
AK014734.1 | - | 6 | 984 | |||
AK080462.1 | - | 6 | 1260 | |||
AK090062.1 | - | 6 | 1271 | |||
BC043112.1 | - | 6 | 1225 | |||
BC072655.1 | - | 6 | 1254 | |||
BC117754.1 | - | 6 | 872 | 40090588 | ||
BC117755.1 | - | 6 | 872 | 40090589 | ||
BC158056.1 | - | 6 | 1023 | 9088164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
