siRNA Details
Gene Symbol: Gm5082 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145878.1 | NP_001139350.1 | 3 | 1030 | ||
NM_177808.4 | NP_808476.2 | 5 | 1193 | |||
GenBank mRNA | AK037195.1 | - | 5 | 1193 | ||
BC098182.1 | - | 3 | 1030 | |||
BC147093.1 | - | 1 | 679 | 9056161 | ||
BC147094.1 | - | 1 | 679 | 9056156 |
