siRNA Details
Gene Symbol: Slc25a48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177809.4 | NP_808477.2 | 6 | 839 | ||
GenBank mRNA | AK054181.1 | - | 6 | 840 | ||
BC107250.1 | - | 6 | 803 | 40052351 | ||
BC107251.1 | - | 6 | 803 | 40052352 | ||
DV649007.1 | - | 3 | 450 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
