siRNA Details
Gene Symbol: Gm5084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037190.1 | - | 3 | 2436 | ||
BC134405.1 | - | 1 | 785 | 40142442 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037190.1 | - | 3 | 2436 | ||
BC134405.1 | - | 1 | 785 | 40142442 |