siRNA Details
Gene Symbol: Npm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181345.3 | NP_851990.2 | 4 | 387 | ||
GenBank mRNA | AK054533.1 | - | 4 | 388 | ||
AK139497.1 | - | 4 | 388 | |||
AY262112.1 | - | 4 | 387 | |||
BC048751.1 | - | 4 | 362 | |||
BC104058.1 | - | 4 | 349 | 40040361 | ||
BC104059.1 | - | 4 | 349 | 40040362 |
