siRNA Details
Gene Symbol: Skint7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142775.1 | NP_001136247.1 | 4 | 747 | ||
NM_177818.3 | NP_808486.2 | 4 | 747 | |||
GenBank mRNA | AK048392.1 | - | 4 | 642 | ||
AK132639.1 | - | 4 | 752 | |||
AV252647.2 | - | 1 | 99 | |||
EF494899.1 | - | 585 | ||||
EF494900.1 | - | 585 | ||||
EU099306.1 | - | 585 |
