siRNA Details
Gene Symbol: Apol10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177820.4 | NP_808488.2 | 4 | 771 | ||
GenBank mRNA | AK033678.1 | - | 743 | |||
AK144113.1 | - | 4 | 775 | |||
BC147348.1 | - | 717 | 9056388 | |||
BC147349.1 | - | 717 | 9056408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
