siRNA Details
Gene Symbol: Hs3st6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012402.1 | NP_001012402.1 | 2 | 897 | ||
GenBank mRNA | AK146974.1 | - | 3 | 1036 | ||
AY574375.1 | - | 2 | 897 | |||
BC132520.1 | - | 2 | 884 | 40130797 | ||
BC138814.1 | - | 2 | 884 | 8861836 | ||
BC145424.1 | - | 2 | 854 | 9053982 | ||
BC145425.1 | - | 3 | 976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
