siRNA Details
Gene Symbol: Mslnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177822.3 | NP_808490.1 | 5 | 674 | ||
GenBank mRNA | AK028891.1 | - | 5 | 674 | ||
BC052484.1 | - | 4 | 410 | 6312790 |

More Information
Other Species Targeted

Gene Ontology Categories:
