siRNA Details
Gene Symbol: Lhfpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026571.2 | NP_080847.2 | 1 | 379 | ||
GenBank mRNA | AK020389.1 | - | 1 | 381 | ||
AK020670.1 | - | 1 | 367 | |||
BC099483.1 | - | 1 | 277 | 30848018 | ||
BC125264.1 | - | 1 | 216 | |||
BC147842.1 | - | 1 | 177 | 9087559 | ||
BC147847.1 | - | 1 | 177 | 9087564 | ||
BY721459.1 | - | 1 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
