siRNA Details
Gene Symbol: A530064D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178796.6 | NP_848911.2 | 3 | 795 | ||
GenBank mRNA | AB372005.1 | - | 3 | 572 | ||
AK080114.1 | - | 3 | 756 | |||
BC139185.1 | - | 3 | 630 | 8862207 | ||
BC139186.1 | - | 3 | 630 | 8862208 | ||
EU003111.1 | - | 3 | 526 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
