siRNA Details
Gene Symbol: Treml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033405.2 | NP_001028577.2 | 2 | 816 | ||
GenBank mRNA | AK013294.1 | - | 2 | 500 | ||
AK037593.1 | - | 2 | 468 | |||
AK132018.1 | - | 3 | 848 | |||
DQ087184.1 | - | 2 | 340 | |||
DQ341272.1 | - | 2 | 915 |
