siRNA Details
Gene Symbol: Spink10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177829.3 | NP_808497.1 | 5 | 401 | ||
GenBank mRNA | AK039062.1 | - | 5 | 402 | ||
AK162840.1 | - | 5 | 369 | |||
BC125360.1 | - | 5 | 317 | 40129875 | ||
BC125362.1 | - | 5 | 317 | 40129877 | ||
DQ437330.1 | - | 5 | 401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
