siRNA Details
Gene Symbol: Zfp532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207255.2 | NP_997138.1 | 3 | 760 | ||
GenBank mRNA | AK083001.1 | - | 3 | 637 | ||
AK129410.1 | - | 4011 | ||||
BC067032.1 | - | 3 | 760 | 6841127 | ||
BC094671.1 | - | 3 | 764 | 30618961 |
