siRNA Details
Gene Symbol: Zfp516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177464.1 | NP_001170935.1 | 3 | 3395 | ||
NM_183033.2 | NP_898854.1 | 3 | 3417 | |||
GenBank mRNA | AK084274.1 | - | 1 | 370 | ||
AK129092.1 | - | 1 | 947 | |||
BC053104.1 | - | 2 | 3208 | 6406544 |
