siRNA Details
Gene Symbol: Gm815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033407.2 | NP_001028579.1 | 4 | 349 | ||
GenBank mRNA | AK136142.1 | - | 4 | 350 | ||
BC147433.1 | - | 4 | 317 | 9056486 | ||
BC172065.1 | - | 4 | 355 | |||
BC172070.1 | - | 4 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
