siRNA Details
Gene Symbol: Dennd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166501.1 | NP_001159973.1 | 8, 9 | 668 | ||
NM_181347.3 | NP_851992.1 | 7, 8 | 534 | |||
GenBank mRNA | AK155727.1 | - | 7, 8 | 535 | ||
BC141388.1 | - | 7, 8 | 530 | 9056001 | ||
BC141389.1 | - | 7, 8 | 530 | 9056002 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
