siRNA Details
Gene Symbol: Rnf224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033410.2 | NP_001028582.1 | 2 | 451 | ||
GenBank mRNA | AK146725.1 | - | 2 | 452 | ||
BC147425.1 | - | 1 | 644 | 9056501 | ||
BC147426.1 | - | 1 | 644 | 9056480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
