siRNA Details
Gene Symbol: Il1bos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076405.1 | - | 3 | 792 | ||
BC111869.1 | - | 1 | 580 | |||
BC112374.1 | - | 1 | 578 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il1bos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076405.1 | - | 3 | 792 | ||
BC111869.1 | - | 1 | 580 | |||
BC112374.1 | - | 1 | 578 |