siRNA Details
Gene Symbol: Nol4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001986.2 | NP_001001986.2 | 6 | 989 | ||
NM_001134300.2 | NP_001127772.1 | 9 | 1774 | |||
GenBank mRNA | AK139459.1 | - | 6 | 989 | ||
AK147524.1 | - | 9 | 1776 | |||
AK157863.1 | - | 6 | 970 | |||
BC075648.1 | - | 8 | 1185 | 30620653 | ||
BC079552.1 | - | 6 | 931 | |||
BC094234.1 | - | 5 | 840 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
