siRNA Details
Gene Symbol: Svs3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173377.2 | NP_775553.1 | 3 | 1128 | ||
GenBank mRNA | AK035235.1 | - | 3 | 1125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
