siRNA Details
Gene Symbol: Fcrl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113238.1 | NP_001106709.1 | 5 | 800 | ||
NM_183222.3 | NP_899045.3 | 6 | 1064 | |||
GenBank mRNA | AK089756.1 | - | 5 | 770 | ||
AY158090.1 | - | 6 | 1061 | |||
AY506558.1 | - | 6 | 1070 | |||
BC137997.1 | - | 6 | 998 | 8861016 | ||
BC137998.1 | - | 6 | 998 | 8861017 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
