siRNA Details
Gene Symbol: Fcrl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183222.3 | NP_899045.3 | 3 | 431 | ||
GenBank mRNA | AY158090.1 | - | 3 | 431 | ||
AY506558.1 | - | 3 | 440 | |||
BC137997.1 | - | 3 | 368 | 8861016 | ||
BC137998.1 | - | 3 | 368 | 8861017 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
