siRNA Details
Gene Symbol: Fam19a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183224.3 | NP_899047.1 | 4, 5 | 698 | ||
GenBank mRNA | AK078681.1 | - | 4, 5 | 699 | ||
AY325122.1 | - | 3, 4 | 300 | |||
BC118531.1 | - | 4, 5 | 388 | 40095917 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
