siRNA Details
Gene Symbol: Fam19a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183224.3 | NP_899047.1 | 7 | 1237 | ||
GenBank mRNA | AK078681.1 | - | 7 | 1239 | ||
BC118531.1 | - | 7 | 927 | 40095917 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
