siRNA Details
Gene Symbol: Dennd4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_184088.1 | NP_908976.1 | 12 | 2070 | ||
GenBank mRNA | AK155220.1 | - | 12 | 2071 | ||
AK158927.1 | - | 2 | 1516 | |||
BC146602.1 | - | 11 | 1695 | 40099465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
